missing translation for 'onlineSavingsMsg'
Learn More

Gallium antimonide, 99.99% (metals basis)

CAS: 12064-03-8

$97.10 - $400.73

Chemical Identifiers

CAS 12064-03-8
MDL Number MFCD00016101
Synonym gallium antimonide, stibinogallane, galliumantimonid, gallium antimonide, polycryst, gallium antimonide, single crystal substrate , <100>, diam. x thickness 2 in. x 0.5 mm
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1293303
View Documents
Thermo Scientific Chemicals
01293303
1 g
Each for $97.10
Only null left
 
AA1293306
View Documents
Thermo Scientific Chemicals
01293306
5 g
Each for $400.73
Only null left
 
Description

Description

Gallium antimonide can be used for Infrared detectors, infrared LEDs and lasers and transistors, and thermophotovoltaic systems. It can be used as a component of photoresists and other composites where high refractive index is desirable

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12064-03-8
MFCD00016101
gallium antimonide, stibinogallane, galliumantimonid, gallium antimonide, polycryst, gallium antimonide, single crystal substrate , <100>, diam. x thickness 2 in. x 0.5 mm

Specifications

710°C
Pieces
MFCD00016101
gallium antimonide, stibinogallane, galliumantimonid, gallium antimonide, polycryst, gallium antimonide, single crystal substrate , <100>, diam. x thickness 2 in. x 0.5 mm
191.47
Odorless
(metals basis)
12064-03-8
1 g
UN1549
Insoluble in water.
99.99%
Gallium antimonide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.

EINECSNumber : 235-058-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only