missing translation for 'onlineSavingsMsg'
Learn More

Gallium, 99.9999%, (trace metal basis)

Catalog No. p-4509683
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC193070010 1 g Plastic bottle
AC193070050 5 g Plastic bottle
AC193070250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC193070010 Supplier Thermo Scientific Chemicals Supplier No. 193070010
Only null left

CAS: 7440-55-3 | Ga | 69.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 7440-55-3
Molecular Formula Ga
Molecular Weight (g/mol) 69.72
MDL Number MFCD00134045
InChI Key GYHNNYVSQQEPJS-UHFFFAOYSA-N
Synonym gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
PubChem CID 5360835
ChEBI CHEBI:49631
IUPAC Name gallium
SMILES [Ga]

Specifications

Color Gray
Melting Point 29.7°C
Boiling Point 2400.0°C
Density 5.9000g/mL
Quantity 1 g
Packaging Plastic bottle
Form Solid and/or Liquid
Assay Percent Range 99.9999%
Linear Formula Ga
Solubility Information Solubility in water: insoluble.
Formula Weight 69.72
Percent Purity 99.9999%
Grade Trace Metal Basis
Chemical Name or Material Gallium
Show More Show Less
Hazard Category Warning
Recommended Storage 4°C

RUO – Research Use Only