missing translation for 'onlineSavingsMsg'
Learn More

Gallic Acid Monohydrate, ACS Reagent, Reagents

Catalog No. 01337591
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337591 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337591 Supplier Labchem Inc Supplier No. C2105500100C2
Only null left

Gallic Acid Monohydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5995-86-8
Molecular Formula C7H5O5
Molecular Weight (g/mol) 169.11
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-M
IUPAC Name 3,4,5-trihydroxybenzoate
SMILES OC1=CC(=CC(O)=C1O)C([O-])=O

Specifications

Melting Point 251°C to 256°C
Color Beige
Grade ACS
Quantity 100 g
Packaging Glass Bottle
Physical Form Solid
Specific Gravity 1.694
Chemical Name or Material Gallic Acid Monohydrate, ACS Reagent