missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallic Acid Monohydrate, ACS Reagent, Reagents
Gallic Acid Monohydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2105500500A5
Specifications
| Gallic Acid Monohydrate, ACS Reagent | |
| 5995-86-8 | |
| C7H5O5 | |
| OC1=CC(=CC(O)=C1O)C([O-])=O | |
| 169.11 | |
| 500 g | |
| Solid |
| 251°C to 256°C | |
| Beige | |
| LNTHITQWFMADLM-UHFFFAOYSA-M | |
| 3,4,5-trihydroxybenzoate | |
| ACS | |
| Poly Bottle | |
| 1.694 |
Chemical Identifiers
| 5995-86-8 | |
| 169.11 | |
| 3,4,5-trihydroxybenzoate |
| C7H5O5 | |
| LNTHITQWFMADLM-UHFFFAOYSA-M | |
| OC1=CC(=CC(O)=C1O)C([O-])=O |