missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallic Acid Monohydrate, ACS Reagent, Reagents
Specifications
Specifications
| Melting Point | 251°C to 256°C |
| Color | Beige |
| Quantity | 25 kg |
| Packaging | Fiber Drum |
| Physical Form | Solid |
| Specific Gravity | 1.694 |
| Grade | ACS |
| Chemical Name or Material | Gallic Acid Monohydrate, ACS Reagent |