missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallic Acid Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 253°C |
| Color | White-Yellow |
| Quantity | 500 g |
| Physical Form | Crystalline Powder |
| Formula Weight | 170.12 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Chemical Name or Material | Gallic Acid Hydrate |
Safety and Handling
| EINECSNumber | (3)-1629&(9)-1351 |
| RTECSNumber | LW7525000 |
| TSCA | Yes |