missing translation for 'onlineSavingsMsg'
Learn More

Gallic acid, 98%

Catalog No. AC410862500
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC410862500 250 g Plastic bottle
AC410860050 5 g Glass Bottle
AC410860010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC410862500 Supplier Thermo Scientific Chemicals Supplier No. 410862500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 149-91-7 | C7H6O5 | 170.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 149-91-7
Molecular Formula C7H6O5
Molecular Weight (g/mol) 170.12
MDL Number MFCD00002510
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-N
Synonym gallic acid, gallate, benzoic acid, 3,4,5-trihydroxy, gallic acid, tech., galop, pyrogallol-5-carboxylic acid, kyselina gallova, 3,4,5-trihydroxybenzoate, ccris 5523, kyselina gallova czech
PubChem CID 370
ChEBI CHEBI:30778
IUPAC Name 3,4,5-trihydroxybenzoic acid
SMILES OC(=O)C1=CC(O)=C(O)C(O)=C1

Specifications

Melting Point 253.0°C
Color Yellow to Beige
Density 1.6940g/mL
Quantity 250 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Powder
Assay Percent Range 98%
Specific Gravity 1.694
Solubility Information Solubility in water: 12g/L cold water. Other solubilities: easily soluble in warmed water, very soluble in ethanol: 160g/L, acetone: 200g/l, soluble in glycerol: 100g/L, slightly soluble in, ether: 10g/L, insoluble in benzene, chloroform, petrol ether
Formula Weight 170.12
Percent Purity 98%
Chemical Name or Material Gallic acid, 98%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-749-9
RTECSNumber LW7525000
TSCA TSCA

RUO – Research Use Only