missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-B Instrument™ Galileo Thermometer
Handcrafted and individually calibrated.
$203.69
| missing translation for 'productCode' | missing translation for 'brand' | Height (English) | Height (Metric) | Subdivision | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| missing translation for 'productCode' | missing translation for 'brand' | Height (English) | Height (Metric) | Subdivision | Price | Quantity & Availability | |||
|
S95612
|
H-B Instrument™
B620000200EMD |
24 in. | 610 mm | 2°C |
Each for $203.69
Case of 10 Each for $2,036.90
|
|
|||
Description
- Multi-colored spheres become more or less buoyant as the temperature of the liquid they are in changes
- Each sphere differs in weight by only 0.002g
- Filled with nonhazardous liquid
- Standard models use optical crystal and are accurate to ±1°F
- Economy models use laboratory-grade glass and are accurate to ±2°F
Specifications
| Glass | |
| Non Hazardous Liquid | |
| 1°C |