missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Galena Mineral Specimen

For use as tool to explain specific gravity to students - EISCO Galena Mineral Specimen is used in classroom identification labs and exercises, correlates well to Next Generation Science Standards Evidence Statements.

$119.60 - $119.60

Specifications

For Use With (Application) Study of specific gravity
Product Type Galena Mineral Study Specimen
Width (English) Approx. 1 in.
Width (Metric) Approx. 3 cm
Packaging Clear labeled and barcoded bag
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S24688
View Documents
Eisco™
ESNG0015PK12
12/Pk.
Pack of 12 Each for $119.60
Only null left
 
Description

Description

  • Far denser than most minerals
  • Reflective cubic crystals offer shining example of metallic luster
  • Comes in clear labeled and barcoded bag for storage
Specifications

Specifications

Study of specific gravity
Approx. 1 in.
Clear labeled and barcoded bag
Galena Mineral Study Specimen
Approx. 3 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'