missing translation for 'onlineSavingsMsg'
Learn More

Galantamine Hydrobromide 98.0+%, TCI America™

Catalog No. p-7130232
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0293100MG 100 mg
G02931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0293100MG Supplier TCI America Supplier No. G0293100MG

Chemical Identifiers

CAS 4-4-1953
Molecular Formula C17H22BrNO3
Molecular Weight (g/mol) 368.27
InChI Key QORVDGQLPPAFRS-XPSHAMGMSA-N
Synonym galanthamine hydrobromide, galantamine hydrobromide, nivalin, razadyne, mj4ptd2vvw, unii-mj4ptd2vvw, nivaline pharmaceutical, galanthamine hbr, lycoremine hydrobromide
PubChem CID 121587
SMILES CN1CCC23C=CC(CC2OC4=C(C=CC(=C34)C1)OC)O.Br

Specifications

Melting Point 256°C
Color White-Yellow
Quantity 100 mg
UN Number 2811
Formula Weight 368.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Galantamine Hydrobromide
RTECSNumber DF8045500
TSCA No
Recommended Storage Freezer