missing translation for 'onlineSavingsMsg'
Learn More

Galantamine Hydrobromide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018334
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018334 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018334 Supplier Merck Emd Millipore Supplier No. PHR1623500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Galantamine Hydrobromide, a reversible, competitive acetyl cholinesterase inhibitor, is suggested for the remedy of mild to moderate dementia of the Alzheimer′s type. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 4-4-1953
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C17H21 NO3 · HBr
MDL Number MFCD26142877
Synonym Galantamine hydrobromide; Galanthamine hydrobromide
Type Certified Reference Material
Quantity 500 mg
Formula Weight 368.27
Grade Certified Reference Material