missing translation for 'onlineSavingsMsg'
Learn More

Gal beta(1-4)Glc-beta-MP 96.0+%, TCI America™

Catalog No. M18051G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M18051G Supplier TCI America Supplier No. M18051G
Only null left

Chemical Identifiers

CAS 150412-80-9
Molecular Formula C19H28O12
Molecular Weight (g/mol) 448.421
InChI Key PJFQMORRZLJWQE-BAGUKLQFSA-N
Synonym 4-Methoxyphenyl 4-O-(beta-D-Galactopyranosyl)-beta-D-glucopyranoside, LacMP
PubChem CID 11453677
IUPAC Name (2S,3R,4S,5R,6R)-2-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)OC3C(C(C(C(O3)CO)O)O)O)O)O

Specifications

Color White
Quantity 1 g
Formula Weight 448.42
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Gal beta(1-4)Glc-beta-MP
TSCA No
Recommended Storage Freezer