missing translation for 'onlineSavingsMsg'
Learn More

Gal beta(1-3)GlcNAc beta(1-3)Gal beta(1-4)Glc-beta-pNP 97.0+%, TCI America™

Catalog No. g03485mg
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03485MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G03485MG Supplier TCI America Supplier No. G03485MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 148705-09-3
Molecular Formula C32H48N2O23
Molecular Weight (g/mol) 828.73
MDL Number MFCD15072170
InChI Key UUSZRTFVRDAQGP-UHFFFAOYNA-N
PubChem CID 44630014
IUPAC Name N-{2-[(2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl)oxy]-5-hydroxy-6-(hydroxymethyl)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-3-yl}acetamide
SMILES CC(=O)NC1C(OC2C(O)C(CO)OC(OC3C(O)C(O)C(OC4=CC=C(C=C4)[N+]([O-])=O)OC3CO)C2O)OC(CO)C(O)C1OC1OC(CO)C(O)C(O)C1O

Specifications

Color White-Yellow
Quantity 5 mg
Formula Weight 828.73
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Gal beta(1-3)GlcNAc beta(1-3)Gal beta(1-4)Glc-beta-pNP
TSCA No
Recommended Storage Freezer