missing translation for 'onlineSavingsMsg'
Learn More

Gal beta(1-3)GalNAc-alpha-pNP 97.0+%, TCI America™

Catalog No. g03755mg
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03755MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G03755MG Supplier TCI America Supplier No. G03755MG

Chemical Identifiers

CAS 59837-14-8
Molecular Formula C20H28N2O13
Molecular Weight (g/mol) 504.445
MDL Number MFCD00064051
InChI Key INMOOBMAIAWVBW-UVFFFCKLSA-N
PubChem CID 11168041
IUPAC Name N-[(2R,3R,4R,5R,6R)-5-hydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)-4-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-3-yl]acetamide
SMILES CC(=O)NC1C(C(C(OC1OC2=CC=C(C=C2)[N+](=O)[O-])CO)O)OC3C(C(C(C(O3)CO)O)O)O

Specifications

Melting Point 228°C
Color White
Quantity 5 mg
Formula Weight 504.45
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Gal beta(1-3)GalNAc-alpha-pNP
TSCA No
Recommended Storage Freezer