missing translation for 'onlineSavingsMsg'
Learn More

Gal[26Bn]beta(1-4)Glc[236Bn]-beta-MP 98.0+%, TCI America™

Catalog No. m16861g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16861G 1 g
M16865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16861G Supplier TCI America Supplier No. M16861G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 358681-61-5 | C54H58O12 | 899.046 g/mol

Chemical Identifiers

CAS 358681-61-5
Molecular Formula C54H58O12
Molecular Weight (g/mol) 899.046
InChI Key XEEOIRLVLCPQPN-HGKZHNLFSA-N
PubChem CID 11158821
IUPAC Name (2R,3R,4S,5R,6S)-6-[(2R,3R,4S,5R,6S)-6-(4-methoxyphenoxy)-4,5-bis(phenylmethoxy)-2-(phenylmethoxymethyl)oxan-3-yl]oxy-5-phenylmethoxy-2-(phenylmethoxymethyl)oxane-3,4-diol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)COCC3=CC=CC=C3)OC4C(C(C(C(O4)COCC5=CC=CC=C5)O)O)OCC6=CC=CC=C6)OCC7=CC=CC=C7)OCC8=CC=CC=C8

Specifications

Melting Point 130°C
Color White
Quantity 1 g
Synonym 4-Methoxyphenyl 4-O-(2,6-Di-O-benzyl-beta-D-galactopyranosyl)-2,3,6-tri-O-benzyl-beta-D-glucopyranoside, LacMP 3′C4′C-OH, per OBn
Formula Weight 899.05
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Gal[26Bn]beta(1-4)Glc[236Bn]-beta-MP
TSCA No
Recommended Storage Refrigerator