missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gal[236Bn]beta(1-4)Glc[236Bn]-beta-MP 92.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| Synonym | LacMP 4′C-OH, per OBn, 4-Methoxyphenyl 4-O-(2,3,6-Tri-O-benzyl-beta-D-galactopyranosyl)-2,3,6-tri-O-benzyl-beta-D-glucopyranoside |
| Formula Weight | 989.17 |
| Percent Purity | ≥92.0% (HPLC) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Gal[236Bn]beta(1-4)Glc[236Bn]-beta-MP |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |