missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium pieces, 20mm (0.79in) & down, 99.9% (REO)

Catalog No. aa0011809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0011809 10 g
AA0011818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0011809 Supplier Thermo Scientific Chemicals Supplier No. 00011809
Only null left

CAS: 7440-54-2 | Gd | 157.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon

Chemical Identifiers

CAS 7440-54-2
Molecular Formula Gd
Molecular Weight (g/mol) 157.25
MDL Number MFCD00011022
InChI Key UIWYJDYFSGRHKR-UHFFFAOYSA-N
Synonym atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
PubChem CID 23982
ChEBI CHEBI:33375
IUPAC Name gadolinium
SMILES [Gd]

Specifications

Color Silver
Physical Form Pieces
Quantity 10 g
UN Number UN3178
Percent Purity 99.9%
Packaging Glass bottle
Assay (REO)
Chemical Name or Material Gadolinium pieces
Hazard Category H228-H260
Hazard Statement GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber 231-162-2
RTECSNumber LW3850000
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only