missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium pieces, 20mm (0.79in) & down, 99.9% (REO)

Catalog No. AA0011818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0011818 50 g
AA0011809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0011818 Supplier Thermo Scientific Chemicals Supplier No. 0011818

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-54-2 | Gd | 157.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon

Chemical Identifiers

CAS 7440-54-2
Molecular Formula Gd
Molecular Weight (g/mol) 157.25
MDL Number MFCD00011022
InChI Key UIWYJDYFSGRHKR-UHFFFAOYSA-N
Synonym atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
PubChem CID 23982
ChEBI CHEBI:33375
IUPAC Name gadolinium
SMILES [Gd]

Specifications

Physical Form Pieces
Quantity 50 g
Packaging Packaged under argon in glass bottle
Assay Percent Range 99.9%
Chemical Name or Material Gadolinium, Rare Earth Oxides Content
Hazard Category H228-H260
Hazard Statement Keep container tightly closed and dry. This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 231-162-2
RTECSNumber LW3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only