missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Gadolinium ingot, 99.9% (REM)

Catalog No. aa45529kg
Change view
Click to view available options
:
1pc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA45529KG 1pc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45529KG Supplier Thermo Scientific™ Supplier No. 45529KG
Only null left

CAS: 7440-54-2 | Gd | 157.25 g/mol

Chemical Identifiers

CAS 7440-54-2
Molecular Formula Gd
Molecular Weight (g/mol) 157.25
MDL Number MFCD00011022
InChI Key UIWYJDYFSGRHKR-UHFFFAOYSA-N
IUPAC Name gadolinium
SMILES [Gd]

Specifications

Quantity 1 Pc.
Form Solid
Percent Purity 99.9%
ShelfLife 5 Years
EINECSNumber 231-162-2