missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) perchlorate hexahydrate, 50% w/w aq. soln., Reagent Grade

Catalog No. AA40571NA
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40571NA 2 g
AA40571NC 10 g
AA40571NE 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA40571NA Supplier Thermo Scientific Chemicals Supplier No. 040571NA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14017-52-8

[Gd(DO3A-Pr-ATP)(H2O)2] was synthesized by the reaction of DO3A-Pr-ATP with gadolinium(iii) perchlorate hydrate in water. Gadolinium(III) chelates approved by FDA for human imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14017-52-8
MDL Number MFCD00150239
Synonym gadolinium perchlorate, gadolinium iii perchlorate, acmc-20aku7, gadolinium iii perchlorate solution, gadolinium 3+ triperchlorate, gadolinium perchlorate, crystalline hydrate,, gadolinium iii perchlorate solution, 40 wt. % in h2o

Specifications

Physical Form Liquid
Quantity 2 g
Odor Odorless
UN Number UN3211
Solubility Information Insoluble in water.
Formula Weight 563.69 (455.60 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Chemical Name or Material Gadolinium(III) perchlorate hexahydrate
Hazard Category H271-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
EINECSNumber 237-837-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only