missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) nitride, 99.5% (REO)

Catalog No. p-7080187
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4670903 1 g
AA4670906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4670903 Supplier Thermo Scientific Chemicals Supplier No. 04670903
Only null left

CAS: 25764-15-2 | GdN | 171.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25764-15-2
Molecular Formula GdN
Molecular Weight (g/mol) 171.26
MDL Number MFCD00049943
InChI Key FLATXDRVRRDFBZ-UHFFFAOYSA-N
Synonym gadolinium nitride, gadolinium iii nitride, gadoliniumylidyneamine
PubChem CID 117632
IUPAC Name azanylidynegadolinium
SMILES N#[Gd]

Specifications

Color Black
Physical Form Powder
Quantity 1 g
Formula Weight 171.26
Percent Purity 99.5%
Assay (REO)
Chemical Name or Material Gadolinium (III) nitride
EINECSNumber 247-250-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only