missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) 2,4-pentanedionate hydrate, REacton™, 99.9% (REO)

CAS: 64438-54-6 | C15H21GdO6 | 454.58 g/mol

$124.29 - $434.26

Chemical Identifiers

CAS 64438-54-6
Molecular Formula C15H21GdO6
Molecular Weight (g/mol) 454.58
MDL Number MFCD00013493
InChI Key PJCXQIZIMGZZIT-UHFFFAOYSA-N
Synonym gadolinium iii 2,4-pentanedionate hydrate, reacton, r reo
PubChem CID 131675701
IUPAC Name gadolinium(3+);(Z)-4-oxopent-2-en-2-olate;hydrate
SMILES [Gd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1320406
View Documents
Thermo Scientific Chemicals
01320406
5 g N/A
Only null left
 
AA1320414
View Documents
Thermo Scientific Chemicals
01320414
25 g N/A
Only null left
 
Description

Description

It is used as a laboratory chemicals & for manufacture of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

64438-54-6
454.58
PJCXQIZIMGZZIT-UHFFFAOYSA-N
131675701
[Gd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
C15H21GdO6
MFCD00013493
gadolinium iii 2,4-pentanedionate hydrate, reacton, r reo
gadolinium(3+);(Z)-4-oxopent-2-en-2-olate;hydrate

Specifications

64438-54-6
5 g
MFCD00013493
Insoluble in water.
[Gd+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
454.58
454.58 (Anhydrous)
Reacton
Gadolinium(III) 2,4-pentanedionate hydrate
Powder
C15H21GdO6
gadolinium iii 2,4-pentanedionate hydrate, reacton, r reo
PJCXQIZIMGZZIT-UHFFFAOYSA-N
gadolinium(3+);(Z)-4-oxopent-2-en-2-olate;hydrate
131675701
99.9%
Hygroscopic
(REO)
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only