Learn More
Eisco™ Gabbro Specimen
Gabbro forms when iron-rich magma too heavy to reach surface cools slowly over time.
Supplier:  Eisco™ ESNG0043
 View more versions of this product
Catalog No. S24756
Only null left
Description
Description
  • Plutonic mafic rock forms from iron-rich magma chambers deep in earth's crust
  • For use in comparing densities between rock samples
  • Example of phaneritic mafic rock
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Chemical Identifiers
Igneous Rock, Gabbro
Specifications
Igneous Rock, Gabbro
Clear labeled and barcoded bag
2 to 3 cm
Ideal for comparing densities between rock samples
1 Ea.
Product Suggestions
Product Suggestions