missing translation for 'onlineSavingsMsg'
Learn More

G418 Sulfate (White to Off-white Powder), Fisher BioReagents

Catalog No. BP6735
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP6735 5 g Amber Glass
BP6731 1 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP6735 Supplier Fisher BioReagents Supplier No. FLBP673 5
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108321-42-2
Molecular Formula C20H44N4O18S2
Molecular Weight (g/mol) 692.70
MDL Number MFCD00058314
InChI Key UHEPSJJJMTWUCP-KETIWVBHNA-N
PubChem CID 134129582
IUPAC Name (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid)
SMILES OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O

Specifications

Type G418 Sulfate
Physical Form Solid
Absorbance < 0.015 at 280nm, < 0.10 at 570nm
Packaging Amber Glass
Potency > 730μg/mg
Water 10% Max.
Color White
Quantity 5 g
Assay Percent Range > 98 %
Formula Weight 692.7
Percent Purity >98%
Chemical Name or Material G418 Sulfate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
May impair fertility. May cause skin, eye, and respiratory tract irritation. May cause allergic respiratory and skin reaction. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Use personal protective equipment. Do not breathe vapors/dust. Do not ingest. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:1
Instability:0
Recommended Storage -20°C