missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ G418 Sulfate, Powder
Specifications
Specifications
| Content And Storage | 2° to 8°C |
| Form | Powder |
| Product Type | Antibiotic |
| Quantity | 5 g |
| Content And Storage | 2° to 8°C |
| Form | Powder |
| Product Type | Antibiotic |
| Quantity | 5 g |