missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G418 Disulfate 90.0+%, TCI America™
$313.28 - $1,163.67
Chemical Identifiers
| CAS | 108321-42-2 |
|---|---|
| Molecular Formula | C20H44N4O18S2 |
| Molecular Weight (g/mol) | 692.70 |
| MDL Number | MFCD00058314 |
| InChI Key | UHEPSJJJMTWUCP-KETIWVBHNA-N |
| Synonym | g418 disulfate |
| PubChem CID | 134129582 |
| IUPAC Name | (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid) |
| SMILES | OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O |
Chemical Identifiers
| 108321-42-2 | |
| 692.70 | |
| UHEPSJJJMTWUCP-KETIWVBHNA-N | |
| 134129582 | |
| OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O |
| C20H44N4O18S2 | |
| MFCD00058314 | |
| g418 disulfate | |
| (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid) |
Specifications
| G418 Disulfate | |
| 108321-42-2 | |
| C20H44N4O18S2 | |
| g418 disulfate | |
| OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O | |
| 692.70 | |
| 692.70 | |
| G418 Disulfate |
| Crystalline Powder | |
| White/Yellow | |
| MFCD00058314 | |
| UHEPSJJJMTWUCP-KETIWVBHNA-N | |
| (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid) | |
| 134129582 | |
| ≥90.0% (HPLC,N) |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator