missing translation for 'onlineSavingsMsg'
Learn More

G418 disulfate, Cell Culture Reagent

Catalog No. AAJ6267103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6267103 1 g
AAJ6267106 5 g
AAJ6267114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6267103 Supplier Thermo Scientific Chemicals Supplier No. J6267103

G418 sulfate, geneticin, CAS # 108321-42-2, is an antibiotic closely related to Gentamicin B, the common aminoglycoside antibiotic. | CAS: 108321-42-2 | C20H44N4O18S2 | 692.70 g/mol

G418 disulfate is an aminoglycoside that is toxic to bacteria, yeast, protozoans and helminths. It prevents protein synthesis by blocking the elongation step in prokaryotic and eukaryotic ribosomes. It is used as a selection agent for G418 resistant bacteria or fungi after transformation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• G418 sulfate (geneticin) is an aminoglycoside antibiotic isolated from Micromonospora rhodorangea
• G418 is a bacteriostatic that prevents polypeptide formation by inhibiting the elongation step of polypeptide synthesis
Applications
• G418 sulfate can be used for selection of genetically engineered cells in both eukaryotes and prokaryotes
• Laboratory grade for applications such as biochemistry

Chemical Identifiers

CAS 108321-42-2
Molecular Formula C20H44N4O18S2
Molecular Weight (g/mol) 692.70
MDL Number MFCD00058314
InChI Key UHEPSJJJMTWUCP-KETIWVBHNA-N
Synonym Geneticin disulfate
PubChem CID 134129582
SMILES OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O

Specifications

Type G418 Disulfate
Physical Form Powder
Color White
Quantity 1 g
Solubility Information Soluble in water.
IUPAC Name (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid)
Formula Weight 692.71
Grade Cell Culture
Chemical Name or Material G418 disulfate
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only