missing translation for 'onlineSavingsMsg'
Learn More

G418 disulfate, 50mg/ml solution

Catalog No. aaj63871ad
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63871AD 50 mL
AAJ63871AB 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63871AD Supplier Thermo Scientific Chemicals Supplier No. J63871AD

G418 sulfate, geneticin, CAS # 108321-42-2, is an antibiotic closely related to Gentamicin B, the common aminoglycoside antibiotic. | CAS: 108321-42-2 | C20H44N4O18S2 | 692.70 g/mol

G418 disulfate is used for selection and maintenance of prokaryotic and eukaryotic cells transfected with a iNOS promoter construct and neomycin resistance gene. Optimal concentrations for selection and maintenance vary by cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • G418 sulfate (geneticin) is an aminoglycoside antibiotic isolated from Micromonospora rhodorangea
  • G418 is bacteriostatic that prevents polypeptide formation by inhibiting the elongation step of polypeptide synthesis

Applications

  • G418 sulfate can be used for the selection of genetically engineered cells in both eukaryotes and prokaryotes
  • Laboratory-grade for applications such as biochemistry

Chemical Identifiers

CAS 108321-42-2
Molecular Formula C20H44N4O18S2
Molecular Weight (g/mol) 692.70
MDL Number MFCD00058314
InChI Key UHEPSJJJMTWUCP-KETIWVBHNA-N
Synonym Geneticin
PubChem CID 134129582
IUPAC Name (2R,3S,4R,5R,6S)-5-amino-6-{[(1R,2S,3S,4R,6S)-4,6-diamino-3-{[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy}-2-hydroxycyclohexyl]oxy}-2-(1-hydroxyethyl)oxane-3,4-diol; bis(sulfuric acid)
SMILES OS(O)(=O)=O.OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@@H](O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](C(C)O)[C@@H](O)[C@H](O)[C@H]3N)[C@@H]2O)OC[C@]1(C)O

Specifications

Type G418 Disulfate
Physical Form Liquid
Concentration 50mg/ml solution
Quantity 50 mL
Solubility Information Difficult to mix.
Formula Weight 692.71
Chemical Name or Material G418 disulfate
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only