Wilson Wolf Manufacturing G-Rex 24 Well Plate, 8ml fill volume, 1 each
Catalog No. NC1000861
Each well can expand 1 million cells into between 40 and 80 million cells in about 10 days with only one exchange of medium. Individual Well Specifications: 2cm2gas permeable membrane surface area with 8 ml fill volume.
Catalog No.
NC1000861
Supplier
Wilson Wolf ManufacturingSupplier No.
80192M
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Each well can expand 1 million cells into between 40 and 80 million cells in about 10 days with only one exchange of medium. Individual Well Specifications: 2cm2gas permeable membrane surface area with 8 ml fill volume.