Learn More
Pyramex™ Fyxate™ Safety Eyewear
Description
Specifications
Specifications
| Lens Type | H2MAX Anti-fog |
| Certifications/Compliance | ANSI Z87.1,CSA Z94.3 |
| Lens Coating | Anti-fog |
| Description | Dielectric with Foam padding and quick release strap |
| Lens Tint | Clear |
| Frame Color | Black |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |