Learn More
Electron Microscopy Sciences Quartz Microscope Slides, 50 x 25 mm
Catalog No. 50949205
Encompass
There are many applications that require slides and coverslips that have absolute UV transparency, and high temperature properties. EMS now is introducing the highest quality, clear fused quartz microscope slides and coverslips to meet all of these requirements. Fused quartz microscope slides and coverslips with a window of highest transparency to UV is between 0.15 to 5. Typical flatness measured 2 to 4 bands per inch and soften point is about 1250 degree C.
Size: 50 x 25 mm
Thickness: 1 mm
Catalog No. 50949205 Supplier Electron Microscopy Sciences Supplier No. 7225002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
There are many applications that require slides and coverslips that have absolute UV transparency, and high temperature properties. EMS now is introducing the highest quality, clear fused quartz microscope slides and coverslips to meet all of these requirements. Fused quartz microscope slides and coverslips with a window of highest transparency to UV is between 0.15 to 5. Typical flatness measured 2 to 4 bands per inch and soften point is about 1250 degree C.
Size: 50 x 25 mm
Thickness: 1 mm