Learn More
Perkin Elmer Canada Polyimide Uncoated Deactivated Fused Silica Capillary Tubing,Length: 5.0m, Inner Diameter 0.32mm
Catalog No. 509045821
Encompass
Deactivated Tubing is perfect for making guard columns and retention gaps. Use Splitters(p/n N9303448) and connectors (p/n N9302149) to attach tubing to polymide coated columns.
Catalog No. 509045821 Supplier Perkin Elmer Canada Supplier No. N9301357
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Deactivated Tubing is perfect for making guard columns and retention gaps. Use Splitters(p/n N9303448) and connectors (p/n N9302149) to attach tubing to polymide coated columns.