missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furoin 96.0+%, TCI America™
$194.44 - $608.36
Chemical Identifiers
| CAS | 552-86-3 |
|---|---|
| Molecular Formula | C10H8O4 |
| Molecular Weight (g/mol) | 192.17 |
| MDL Number | MFCD00003246 |
| InChI Key | MIJRFWVFNKQQDK-UHFFFAOYNA-N |
| Synonym | furoin, 2,2'-furoin, 1,2-di furan-2-yl-2-hydroxyethanone, furoylfurylcarbinol, ethanone, 1,2-di-2-furanyl-2-hydroxy, alpha-furoin, .alpha.-furoin, ethanone, 1,2-di-2-furyl-2-hydroxy, 1,2-bis furan-2-yl-2-hydroxyethan-1-one, 1,2-di-2-furanyl-2-hydroxyethanone |
| PubChem CID | 11100 |
| IUPAC Name | 1,2-bis(furan-2-yl)-2-hydroxyethan-1-one |
| SMILES | OC(C(=O)C1=CC=CO1)C1=CC=CO1 |
Chemical Identifiers
| 552-86-3 | |
| 192.17 | |
| MIJRFWVFNKQQDK-UHFFFAOYNA-N | |
| 11100 | |
| OC(C(=O)C1=CC=CO1)C1=CC=CO1 |
| C10H8O4 | |
| MFCD00003246 | |
| furoin, 2,2'-furoin, 1,2-di furan-2-yl-2-hydroxyethanone, furoylfurylcarbinol, ethanone, 1,2-di-2-furanyl-2-hydroxy, alpha-furoin, .alpha.-furoin, ethanone, 1,2-di-2-furyl-2-hydroxy, 1,2-bis furan-2-yl-2-hydroxyethan-1-one, 1,2-di-2-furanyl-2-hydroxyethanone | |
| 1,2-bis(furan-2-yl)-2-hydroxyethan-1-one |
Specifications
| 552-86-3 | |
| C10H8O4 | |
| 5 g | |
| MIJRFWVFNKQQDK-UHFFFAOYNA-N | |
| 1,2-bis(furan-2-yl)-2-hydroxyethan-1-one | |
| 11100 | |
| ≥96.0% (GC) | |
| Furoin |
| 139°C | |
| MFCD00003246 | |
| furoin, 2,2'-furoin, 1,2-di furan-2-yl-2-hydroxyethanone, furoylfurylcarbinol, ethanone, 1,2-di-2-furanyl-2-hydroxy, alpha-furoin, .alpha.-furoin, ethanone, 1,2-di-2-furyl-2-hydroxy, 1,2-bis furan-2-yl-2-hydroxyethan-1-one, 1,2-di-2-furanyl-2-hydroxyethanone | |
| OC(C(=O)C1=CC=CO1)C1=CC=CO1 | |
| 192.17 | |
| 192.17 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : KM5774095
TSCA : Yes