missing translation for 'onlineSavingsMsg'
Learn More

Furoin 96.0+%, TCI America™

Catalog No. F00825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F00825G 5 g
F008225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F00825G Supplier TCI America Supplier No. F00825G

Chemical Identifiers

CAS 552-86-3
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00003246
InChI Key MIJRFWVFNKQQDK-UHFFFAOYNA-N
Synonym furoin, 2,2'-furoin, 1,2-di furan-2-yl-2-hydroxyethanone, furoylfurylcarbinol, ethanone, 1,2-di-2-furanyl-2-hydroxy, alpha-furoin, .alpha.-furoin, ethanone, 1,2-di-2-furyl-2-hydroxy, 1,2-bis furan-2-yl-2-hydroxyethan-1-one, 1,2-di-2-furanyl-2-hydroxyethanone
PubChem CID 11100
IUPAC Name 1,2-bis(furan-2-yl)-2-hydroxyethan-1-one
SMILES OC(C(=O)C1=CC=CO1)C1=CC=CO1

Specifications

Melting Point 139°C
Quantity 5 g
Formula Weight 192.17
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Furoin
RTECSNumber KM5774095
TSCA Yes