missing translation for 'onlineSavingsMsg'
Learn More

Furo[3,2-b]pyridine, 95%

Catalog No. AAH3296606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3296606 5 g
AAH3296603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3296606 Supplier Thermo Scientific Chemicals Supplier No. H3296606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 272-62-8 | C7H5NO | 119.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 272-62-8
Molecular Formula C7H5NO
Molecular Weight (g/mol) 119.12
MDL Number MFCD11870744
InChI Key YRTCKZIKGWZNCU-UHFFFAOYSA-N
Synonym furo 3,2-b pyridine, 4-azabenzo b furan, furopyridine, furo 3,2-b pyridin, 4-aza-1-benzofuran
PubChem CID 12210217
IUPAC Name furo[3,2-b]pyridine
SMILES O1C=CC2=NC=CC=C12

Specifications

Boiling Point 79°C to 80°C (12 mmHg)
Refractive Index 1.5753
Quantity 5 g
Formula Weight 119.12
Percent Purity 98%
Chemical Name or Material Furo[3,2-b]pyridine
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only