missing translation for 'onlineSavingsMsg'
Learn More

Furo[3,2-b]pyridine-2-carboxylic acid, 97%

Catalog No. p-7032787
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34400MD 250 mg
AAH3440003 1 g
AAH3440006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH34400MD Supplier Thermo Scientific Chemicals Supplier No. H34400MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 112372-14-2 | C8H5NO3 | 163.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112372-14-2
Molecular Formula C8H5NO3
Molecular Weight (g/mol) 163.13
MDL Number MFCD11588128
InChI Key VHRCHODSRLMXFM-UHFFFAOYSA-N
Synonym furo 3,2-b pyridine-2-carboxylic acid, acmc-20mg3q, 2-carboxyfuro 3,2-b pyridine
PubChem CID 13803071
IUPAC Name furo[3,2-b]pyridine-2-carboxylic acid
SMILES OC(=O)C1=CC2=NC=CC=C2O1

Specifications

Melting Point >300°C
Quantity 250 mg
Formula Weight 163.13
Percent Purity 97%
Chemical Name or Material Furo[3,2-b]pyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only