missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furfurylamine 98.0+%, TCI America™
Supplier: TCI America F0091100ML
Specifications
| Furfurylamine | |
| -70°C | |
| 146°C | |
| MFCD00003258 | |
| 2526 | |
| DDRPCXLAQZKBJP-UHFFFAOYSA-N | |
| furan-2-ylmethanamine | |
| 3438 | |
| ≥98.0% (GC,T) |
| 617-89-0 | |
| Yellow | |
| C5H7NO | |
| 100 mL | |
| furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl | |
| C1=COC(=C1)CN | |
| 97.117 | |
| 97.12 | |
| Liquid |
Chemical Identifiers
| 617-89-0 | |
| 97.117 | |
| DDRPCXLAQZKBJP-UHFFFAOYSA-N | |
| 3438 | |
| C1=COC(=C1)CN |
| C5H7NO | |
| MFCD00003258 | |
| furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl | |
| furan-2-ylmethanamine |
Safety and Handling
EINECSNumber : (9)-1264
RTECSNumber : LU9275000
TSCA : Yes