missing translation for 'onlineSavingsMsg'
Learn More

Furfurylamine 98.0+%, TCI America™

Catalog No. F0091500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0091500ML 500 mL
F009125ML 25 mL
F0091100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F0091500ML Supplier TCI America Supplier No. F0091500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 617-89-0
Molecular Formula C5H7NO
Molecular Weight (g/mol) 97.117
MDL Number MFCD00003258
InChI Key DDRPCXLAQZKBJP-UHFFFAOYSA-N
Synonym furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl
PubChem CID 3438
IUPAC Name furan-2-ylmethanamine
SMILES C1=COC(=C1)CN

Specifications

Melting Point -70°C
Color Yellow
Boiling Point 146°C
Quantity 500 mL
UN Number 2526
Formula Weight 97.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Furfurylamine
EINECSNumber (9)-1264
RTECSNumber LU9275000
TSCA Yes