missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl Methacrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. M13355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13355G 5 g
M133525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13355G Supplier TCI America Supplier No. M13355G
Only null left

Chemical Identifiers

CAS 3454-28-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00046113
InChI Key DWXAVNJYFLGAEF-UHFFFAOYSA-N
Synonym Methacrylic Acid Furfuryl Ester
PubChem CID 76993
IUPAC Name furan-2-ylmethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC1=CC=CO1

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Furfuryl Methacrylate (stabilized with MEHQ)