missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl isocyanate, 97%

Catalog No. AC456490010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456490010 1 g
AC456490050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456490010 Supplier Thermo Scientific Chemicals Supplier No. 456490010

CAS: 71189-15-6 | C6H5NO2 | 123.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71189-15-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
MDL Number MFCD03180268
InChI Key UIADMYLYGJYUSQ-UHFFFAOYSA-N
Synonym 2-isocyanatomethyl furan, furfuryl isocyanate, 2-furylmethyl isocyanate, furfurylisocyanate, furylmethylisocyanate, 2-isocyanatomethyl-furan, 2-furyl methyl isocyanate, furan, 2-isocyanatomethyl
PubChem CID 4223811
IUPAC Name 2-(isocyanatomethyl)furan
SMILES O=C=NCC1=CC=CO1

Specifications

Melting Point 157.0°C to 158.0°C
Density 1.1480g/mL
Flash Point 54.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.148
Formula Weight 123.11
Percent Purity 97%
Chemical Name or Material Furfuryl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause allergy or asthma symptoms or
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and k

RUO – Research Use Only