missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl Acetate 97.0+%, TCI America™

Catalog No. A090325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A090325ML 25 mL
A0903100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A090325ML Supplier TCI America Supplier No. A090325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 623-17-6
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00003251
InChI Key CKOYRRWBOKMNRG-UHFFFAOYSA-N
Synonym furfuryl acetate, 2-furanmethanol, acetate, 2-furanmethanol acetate, 2-acetoxymethylfuran, furfuryl alcohol, acetate, acetic acid furfurylester, acetic acid furfuryl ester, 2-furanmethyl acetate, 2-furfuryl acetate, 2-furylmethyl acetate
PubChem CID 12170
IUPAC Name furan-2-ylmethyl acetate
SMILES CC(=O)OCC1=CC=CO1

Specifications

Color Yellow
Boiling Point 177°C
Quantity 25 mL
Formula Weight 140.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Furfuryl Acetate
EINECSNumber (5)-0032
RTECSNumber LU9120000
TSCA Yes