missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl acetate, 97%

Catalog No. AAA1528522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1528522 100 g
AAA1528514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1528522 Supplier Thermo Scientific Chemicals Supplier No. A1528522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 623-17-6 | C7H8O3 | 140.138 g/mol

Furfuryl acetate is a flavoring ingredient. It was used in the synthesis of 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2-vinylfuran via Vilsmeier-Haack and Wittig reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfuryl acetate is a flavoring ingredient. It was used in the synthesis of 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2-vinylfuran via Vilsmeier-Haack and Wittig reactions.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 623-17-6
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00003251
InChI Key CKOYRRWBOKMNRG-UHFFFAOYSA-N
Synonym furfuryl acetate, 2-furanmethanol, acetate, 2-furanmethanol acetate, 2-acetoxymethylfuran, furfuryl alcohol, acetate, acetic acid furfurylester, acetic acid furfuryl ester, 2-furanmethyl acetate, 2-furfuryl acetate, 2-furylmethyl acetate
PubChem CID 12170
IUPAC Name furan-2-ylmethyl acetate
SMILES CC(=O)OCC1=CC=CO1

Specifications

Density 1.118
Boiling Point 175°C to 177°C
Flash Point 65°C (149°F)
Refractive Index 1.463
Quantity 100 g
Beilstein 116128
Solubility Information Not miscible in water.
Formula Weight 140.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Furfuryl acetate
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement

H227

Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-775-9
RTECSNumber LU9120000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only