missing translation for 'onlineSavingsMsg'
Learn More

Furfural 98.0+%, TCI America™

Catalog No. p-7129481
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F007325G 25 g
F0073500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F007325G Supplier TCI America Supplier No. F007325G
Only null left

Chemical Identifiers

CAS 98-01-1
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.09
MDL Number MFCD00003229
InChI Key HYBBIBNJHNGZAN-UHFFFAOYSA-N
Synonym furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
PubChem CID 7362
ChEBI CHEBI:34768
IUPAC Name furan-2-carbaldehyde
SMILES O=CC1=CC=CO1

Specifications

Melting Point -37°C
Color Yellow
pH >3.0
Boiling Point 160°C
Quantity 25 g
UN Number 1199
Formula Weight 96.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Furfural
EINECSNumber (5)-0040
RTECSNumber LT7000000
TSCA Yes