Learn More
Furazolidone, 98%
CAS: 67-45-8 | C8H7N3O5 | 225.16 g/mol
Supplier: Thermo Scientific Chemicals B2083422
Description
Monoamine oxidase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Furazolidone | |
| 254°C to 256°C (decomposition) | |
| MFCD00010550 | |
| Light sensitive | |
| PLHJDBGFXBMTGZ-WEVVVXLNSA-N | |
| 3-[(E)-(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one | |
| 5323714 | |
| 98% |
| 67-45-8 | |
| C8H7N3O5 | |
| 100 g | |
| furazolidone, furoxone, nitrofurazolidone, furazolidine, nitrofuroxon, furazolidon, diafuron, furaxone, furazol, furazon | |
| C1COC(=O)N1N=CC2=CC=C(O2)[N+](=O)[O-] | |
| 225.16 | |
| 225.16 |
Chemical Identifiers
| 67-45-8 | |
| 225.16 | |
| PLHJDBGFXBMTGZ-WEVVVXLNSA-N | |
| 5323714 | |
| C1COC(=O)N1N=CC2=CC=C(O2)[N+](=O)[O-] |
| C8H7N3O5 | |
| MFCD00010550 | |
| furazolidone, furoxone, nitrofurazolidone, furazolidine, nitrofuroxon, furazolidon, diafuron, furaxone, furazol, furazon | |
| 3-[(E)-(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one |
Safety and Handling
GHS H Statement
H341
Suspected of causing genetic defects.
P201-P202-P281-P308+P313-P501c
H341
EINECSNumber : 200-653-3
RTECSNumber : RQ3675000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only