missing translation for 'onlineSavingsMsg'
Learn More

Furazolidone, 98%

Catalog No. AAB2083414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2083414 25 g
AAB2083422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2083414 Supplier Thermo Scientific Chemicals Supplier No. B2083414

CAS: 67-45-8 | C8H7N3O5 | 225.16 g/mol

Monoamine oxidase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67-45-8
Molecular Formula C8H7N3O5
Molecular Weight (g/mol) 225.16
MDL Number MFCD00010550
InChI Key PLHJDBGFXBMTGZ-WEVVVXLNSA-N
Synonym furazolidone, furoxone, nitrofurazolidone, furazolidine, nitrofuroxon, furazolidon, diafuron, furaxone, furazol, furazon
PubChem CID 5323714
IUPAC Name 3-[(E)-(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one
SMILES C1COC(=O)N1N=CC2=CC=C(O2)[N+](=O)[O-]

Specifications

Melting Point 254°C to 256°C (decomposition)
Quantity 25 g
Sensitivity Light sensitive
Formula Weight 225.16
Percent Purity 98%
Chemical Name or Material Furazolidone
Hazard Category H341
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 200-653-3
RTECSNumber RQ3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only