missing translation for 'onlineSavingsMsg'
Learn More

Furan, 99+%, stabilized

Catalog No. p-3758058
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC181112500 250 mL Glass bottle
AC181110010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC181112500 Supplier Thermo Scientific Chemicals Supplier No. 181112500

CAS: 110-00-9 | C4H4O | 68.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-00-9 , 128-37-0
Molecular Formula C4H4O
Molecular Weight (g/mol) 68.08
MDL Number MFCD00003222
InChI Key YLQBMQCUIZJEEH-UHFFFAOYSA-N
Synonym divinylene oxide, furfuran, tetrole, oxole, oxacyclopentadiene, furane, 1,4-epoxy-1,3-butadiene, axole, rcra waste number u124, furfurane
PubChem CID 8029
ChEBI CHEBI:35559
IUPAC Name furan
SMILES O1C=CC=C1

Specifications

Melting Point -85.6°C
Color Colorless to Yellow
Density 0.9360g/mL
Boiling Point 31.3°C
Flash Point −35°C
Infrared Spectrum Authentic
Assay Percent Range 0.05% max. (Furfural) (GC)
Packaging Glass bottle
Refractive Index 1.4197 to 1.4217
Quantity 250 mL
UN Number 2389
Beilstein 17, 27
Fieser 07,161; 09,227; 10,187; 13,173
Specific Gravity 0.936
Merck Index 15, 4325
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in alcohol,ether and aceton,hydrolysis in water
Stabilizer 0.025 to 0.04% BHT
Formula Weight 68.06
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Furan
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Harmful if inhaled.
May cause cancer.
Harmful to aquatic life with long
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the
EINECSNumber 203-727-3

RUO – Research Use Only