missing translation for 'onlineSavingsMsg'
Learn More

Furan-3-boronic acid, 97%

Catalog No. AAL1983403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1983403 1 g
AAL1983406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1983403 Supplier Thermo Scientific Chemicals Supplier No. L1983403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 55552-70-0 | C4H5BO3 | 111.89 g/mol

Furan-3-boronic acid use furan-3-boronic acid as our coupling partner in many reactions. A chitosan uricase (UOx)-poly (furan-3-boronic acid)(PFBA)-Pd nanoparticles (PdNPs)/plated Pd (Pd plate)/multiwalled carbon nanotubes (MWCNTs)/Au electrode was prepared for fabricating an amperometric biosensor and a biofuel cell (BFC) of uric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furan-3-boronic acid use furan-3-boronic acid as our coupling partner in many reactions. A chitosan uricase (UOx)-poly (furan-3-boronic acid)(PFBA)-Pd nanoparticles (PdNPs)/plated Pd (Pd plate)/multiwalled carbon nanotubes (MWCNTs)/Au electrode was prepared for fabricating an amperometric biosensor and a biofuel cell (BFC) of uric acid.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 55552-70-0
Molecular Formula C4H5BO3
Molecular Weight (g/mol) 111.89
MDL Number MFCD01319007
InChI Key CYEFKCRAAGLNHW-UHFFFAOYSA-N
Synonym furan-3-boronic acid, 3-furanboronic acid, 3-furylboronic acid, furan-3-yl boronic acid, 3-furanylboronic acid, furan-3-ylboranediol, 3-furan boronic acid, furan-3-yl-3-boronic acid, furan-3-yl-boronic acid, boronic acid, 3-furanyl
PubChem CID 2734358
IUPAC Name (furan-3-yl)boronic acid
SMILES OB(O)C1=COC=C1

Specifications

Melting Point 139°C to 144°C
Quantity 1 g
Beilstein 1635653
Solubility Information Soluble in methanol.
Formula Weight 111.89
Percent Purity 97%
Chemical Name or Material Furan-3-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only