Learn More
Furan-2-boronic acid, 97%
CAS: 13331-23-2 | C4H5BO3 | 111.89 g/mol
$565.69 - $565.69
Chemical Identifiers
| CAS | 13331-23-2 |
|---|---|
| Molecular Formula | C4H5BO3 |
| Molecular Weight (g/mol) | 111.89 |
| MDL Number | MFCD00799544 |
| InChI Key | PZJSZBJLOWMDRG-UHFFFAOYSA-N |
| Synonym | 2-furanboronic acid, furan-2-boronic acid, 2-furylboronic acid, 2-furanylboronic acid, furan-2-ylboranediol, furan 2-boronic acid, 2-furan boronic acid, furan-2-yl boronic acid, boronic acid, furanyl, 2furylboronic acid |
| PubChem CID | 2734357 |
| IUPAC Name | furan-2-ylboronic acid |
| SMILES | B(C1=CC=CO1)(O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC359390100
|
Thermo Scientific Chemicals
359390100 |
10 g | Glass bottle |
Each for $565.69
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13331-23-2 | |
| 111.89 | |
| PZJSZBJLOWMDRG-UHFFFAOYSA-N | |
| 2734357 | |
| B(C1=CC=CO1)(O)O |
| C4H5BO3 | |
| MFCD00799544 | |
| 2-furanboronic acid, furan-2-boronic acid, 2-furylboronic acid, 2-furanylboronic acid, furan-2-ylboranediol, furan 2-boronic acid, 2-furan boronic acid, furan-2-yl boronic acid, boronic acid, furanyl, 2furylboronic acid | |
| furan-2-ylboronic acid |
Specifications
| 13331-23-2 | |
| Authentic | |
| Glass bottle | |
| MFCD00799544 | |
| 2-furanboronic acid, furan-2-boronic acid, 2-furylboronic acid, 2-furanylboronic acid, furan-2-ylboranediol, furan 2-boronic acid, 2-furan boronic acid, furan-2-yl boronic acid, boronic acid, furanyl, 2furylboronic acid | |
| B(C1=CC=CO1)(O)O | |
| 111.89 | |
| 111.89 | |
| Crystalline Powder |
| Beige | |
| 96% min. (HPLC) | |
| C4H5BO3 | |
| 10 g | |
| PZJSZBJLOWMDRG-UHFFFAOYSA-N | |
| furan-2-ylboronic acid | |
| 2734357 | |
| 97% | |
| Furan-2-boronic acid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only