missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FURA 2 pentasodium salt
Description
A very sensitive fluorescent calcium chelator
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Physical Form | Solid |
| Quantity | 1 mg |
| Formula Weight | 751.45 |
| Sensitivity | Light and moisture sensitive |
| Chemical Name or Material | FURA 2 pentasodium salt |
Safety and Handling
| TSCA | No |
| storageNote1 | -30° to -10°C |
| Recommended Storage | Store at -20°C; Store under Nitrogen |
RUO – Research Use Only