missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Fura-2/AM, Calbiochem™,

Catalog No. 3449051SET
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3449051SET 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3449051SET Supplier MilliporeSigma™ Supplier No. 3449051SET
Only null left

Cell-permeable ester form of FURA 2

Chemical Identifiers

CAS 08964-32-5
Molecular Formula C44H47N2O24
Synonym 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methylphenoxy)-ethane-N,N,N'N'-tetraacetic Acid Pentaacetoxymethyl Ester

Specifications

Color Yellow
Quantity 1 Set
Solubility Information Soluble in DSMO
Formula Weight 1001.9
Percent Purity ≥95%
Grade HPLC
Physical Form Solid
Chemical Name or Material Fura-2/AM
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above -20°C (-4°F).